Assay Details
| Gene Symbol: enosf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113128.1 | NP_001106599.1 | 8 - 9 | 598 | 71 | |
| XM_012964234.2 | XP_012819688.1 | 9 - 10 | 1028 | 71 | ||
| XM_012964235.2 | XP_012819689.1 | 9 - 10 | 945 | 71 | ||
| XM_012964236.2 | XP_012819690.1 | 9 - 10 | 919 | 71 | ||
| XM_018094523.1 | XP_017950012.1 | 8 - 9 | 1117 | 71 | ||
| GenBank mRNA | BC155430.1 | - | 8 - 9 | 598 | 71 | |
More Information
Set Membership: |
|

enosf1
Genomic Map