Assay Details
| Gene Symbol: jade2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113130.1 | NP_001106601.1 | 5 - 6 | 865 | 77 | |
| XM_012958513.2 | XP_012813967.1 | 5 - 6 | 2846 | 77 | ||
| XM_012958514.2 | XP_012813968.1 | 6 - 7 | 2930 | 77 | ||
| GenBank mRNA | BC155434.1 | - | 5 - 6 | 865 | 77 | |
More Information
Set Membership: |
|

jade2
Genomic Map