Assay Details
| Gene Symbol: gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113693.1 | NP_001107165.1 | 1 - 2 | 146 | 66 | |
| GenBank mRNA | EF166084.1 | - | 1 - 2 | 146 | 66 | |
More Information
Set Membership: |
|

gnrh1
Genomic Map