Assay Details
Gene Symbol: gnrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113693.1 | NP_001107165.1 | 2 - 3 | 229 | 80 | |
GenBank mRNA | EF166084.1 | - | 2 - 3 | 229 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
