Assay Details
| Gene Symbol: ccdc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113924.1 | NP_001107396.1 | 2 - 3 | 544 | 76 | |
| GenBank mRNA | BC157336.1 | - | 2 - 3 | 544 | 76 | |
| BC171280.1 | - | 2 - 3 | 478 | 76 | ||
| BC171282.1 | - | 2 - 3 | 478 | 76 | ||
| CR855760.2 | - | 2 - 3 | 582 | 76 | ||
More Information
Set Membership: |
|

ccdc59
Genomic Map