Assay Details
Gene Symbol: cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114489.1 | NP_001107961.1 | 6 - 7 | 902 | 74 | |
XM_012971718.2 | XP_012827172.1 | 7 - 8 | 1025 | 74 | ||
XM_018097721.1 | XP_017953210.1 | 5 - 6 | 859 | 74 | ||
GenBank mRNA | BC088607.1 | - | 6 - 7 | 739 | 74 | |
BC118711.1 | - | 6 - 7 | 879 | 74 | ||
BC159128.1 | - | 6 - 7 | 902 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
