Assay Details
| Gene Symbol: sept8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122727.1 | NP_001116199.1 | 5 - 6 | 896 | 64 | |
| GenBank mRNA | BC160437.1 | - | 5 - 6 | 896 | 64 | |
| CU075437.1 | - | 5 - 6 | 729 | 64 | ||
More Information
Set Membership: |
|

sept8
Genomic Map