Assay Details
Gene Symbol: ndnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122800.1 | NP_001116272.1 | 3 - 4 | 575 | 76 | |
XM_012965883.2 | XP_012821337.1 | 3 - 4 | 439 | 76 | ||
GenBank mRNA | BC136060.1 | - | 3 - 4 | 565 | 76 | |
BC152025.1 | - | 3 - 4 | 576 | 76 | ||
BC161015.1 | - | 3 - 4 | 575 | 76 | ||
CR942677.2 | - | 3 - 4 | 568 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
