Assay Details
| Gene Symbol: ism2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123017.1 | NP_001116489.1 | 5 - 6 | 1081 | 80 | |
| XM_012968542.2 | XP_012823996.1 | 5 - 6 | 1103 | 80 | ||
| GenBank mRNA | BC161601.1 | - | 5 - 6 | 1081 | 80 | |
| CR848304.2 | - | 3 - 4 | 421 | 80 | ||
More Information
Set Membership: |
|

ism2
Genomic Map