Assay Details
| Gene Symbol: sfrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123406.1 | NP_001116878.1 | 1 - 2 | 734 | 101 | |
| GenBank mRNA | BC159365.1 | - | 1 - 2 | 734 | 101 | |
| CR926172.2 | - | 1 - 2 | 738 | 101 | ||
More Information
Set Membership: |
|

sfrp1
Genomic Map