Assay Details
| Gene Symbol: dcun1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126565.1 | NP_001120037.1 | 4 - 5 | 582 | 89 | |
| XM_012956436.2 | XP_012811890.1 | 5 - 6 | 709 | 89 | ||
| XM_012956437.2 | XP_012811891.1 | 5 - 6 | 1302 | 89 | ||
| XM_012956438.2 | XP_012811892.1 | 4 - 5 | 1205 | 89 | ||
| GenBank mRNA | BC158344.1 | - | 4 - 5 | 582 | 89 | |
More Information
Set Membership: |
|

dcun1d2
Genomic Map