Assay Details
| Gene Symbol: nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126661.1 | NP_001120133.1 | 2 - 3 | 152 | 87 | |
| GenBank mRNA | BC159063.1 | - | 2 - 3 | 152 | 87 | |
| CT025236.2 | - | 2 - 3 | 105 | 87 | ||
More Information
Set Membership: |
|

nxt2
Genomic Map