Assay Details
| Gene Symbol: fam32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126664.1 | NP_001120136.1 | 2 - 3 | 247 | 88 | |
| GenBank mRNA | BC159080.1 | - | 2 - 3 | 247 | 88 | |
| CT025469.2 | - | 2 - 3 | 243 | 88 | ||
More Information
Set Membership: |
|

fam32a
Genomic Map