Assay Details
| Gene Symbol: tmem59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127135.1 | NP_001120607.1 | 7 - 8 | 836 | 71 | |
| GenBank mRNA | BC161736.1 | - | 7 - 8 | 836 | 71 | |
| CU075434.1 | - | 7 - 8 | 817 | 71 | ||
More Information
Set Membership: |
|

tmem59
Genomic Map