Assay Details
Gene Symbol: wdr83os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126516.1 | NP_001119988.1 | 578 | 102 | ||
GenBank mRNA | BC158245.1 | - | 578 | 102 | ||
CR942720.2 | - | 1 - 2 | 66 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
