Assay Details
| Gene Symbol: fam192a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126775.1 | NP_001120247.1 | 2 - 3 | 209 | 76 | |
| XM_012960609.2 | XP_012816063.1 | 2 - 3 | 261 | 76 | ||
| GenBank mRNA | BC160456.1 | - | 2 - 3 | 209 | 76 | |
More Information
Set Membership: |
|

fam192a
Genomic Map