Assay Details
| Gene Symbol: socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127426.1 | NP_001120898.1 | 2 - 3 | 406 | 77 | |
| XM_012958618.2 | XP_012814072.1 | 2 - 3 | 994 | 77 | ||
| GenBank mRNA | EF544587.1 | - | 2 - 3 | 406 | 77 | |
More Information
Set Membership: |
|

socs2
Genomic Map