Assay Details
| Gene Symbol: ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128648.1 | NP_001122120.1 | 2 - 3 | 329 | 84 | |
| GenBank mRNA | BC118831.1 | - | 2 - 3 | 303 | 84 | |
| BC167664.1 | - | 2 - 3 | 329 | 84 | ||
More Information
Set Membership: |
|

ngrn
Genomic Map