Assay Details
| Gene Symbol: klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129932.1 | NP_001123404.1 | 1 - 2 | 321 | 58 | |
| XM_012964357.1 | XP_012819811.1 | 1 - 2 | 397 | 58 | ||
| XM_018094561.1 | XP_017950050.1 | 1 - 2 | 397 | 58 | ||
| XM_018094562.1 | XP_017950051.1 | 1 - 2 | 397 | 58 | ||
| GenBank mRNA | BC167595.1 | - | 1 - 2 | 321 | 58 | |
More Information
Set Membership: |
|

klhl18
Genomic Map