Assay Details
| Gene Symbol: sesn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129939.1 | NP_001123411.1 | 1 - 2 | 219 | 70 | |
| GenBank mRNA | BC167380.1 | - | 1 - 2 | 219 | 70 | |
More Information
Set Membership: |
|

sesn1
Genomic Map