Assay Details
Gene Symbol: mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130244.1 | NP_001123716.1 | 1 - 2 | 130 | 64 | |
GenBank mRNA | BC167338.1 | - | 1 - 2 | 130 | 64 | |
CU075761.1 | - | 1 - 2 | 115 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
