Assay Details
Gene Symbol: stard3nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130258.1 | NP_001123730.1 | 4 - 5 | 490 | 63 | |
XM_012964364.1 | XP_012819818.1 | 4 - 5 | 668 | 63 | ||
GenBank mRNA | BC167376.1 | - | 4 - 5 | 490 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
