Assay Details
Gene Symbol: akap17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134815.2 | NP_001128287.2 | 2 - 3 | 890 | 74 | |
GenBank mRNA | BC168422.1 | - | 2 - 3 | 844 | 74 | |
CR760259.2 | - | 2 - 3 | 863 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
