Assay Details
Gene Symbol: akap17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134815.2 | NP_001128287.2 | 4 - 5 | 1286 | 69 | |
GenBank mRNA | BC168422.1 | - | 4 - 5 | 1241 | 69 | |
CX419330.1 | - | 2 - 3 | 372 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
