Assay Details
Gene Symbol: jpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142023.1 | NP_001135495.1 | 2 - 3 | 288 | 83 | |
XM_012970172.1 | XP_012825626.1 | 2 - 3 | 397 | 83 | ||
GenBank mRNA | BC167895.1 | - | 2 - 3 | 288 | 83 | |
CU075616.1 | - | 2 - 3 | 267 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
