Assay Details
| Gene Symbol: odam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145745.2 | NP_001139217.1 | 8 - 9 | 704 | 63 | |
| XM_012957733.1 | XP_012813187.1 | 8 - 9 | 704 | 63 | ||
| GenBank mRNA | EU642609.1 | - | 8 - 9 | 705 | 63 | |
More Information
Set Membership: |
|

odam
Genomic Map