Assay Details
| Gene Symbol: jpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145748.1 | NP_001139220.1 | 2 - 3 | 245 | 61 | |
| GenBank mRNA | BC077053.1 | - | 2 - 3 | 238 | 61 | |
| BC161642.1 | - | 2 - 3 | 222 | 61 | ||
More Information
Set Membership: |
|

jpt1
Genomic Map