Assay Details
Gene Symbol: jpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145748.1 | NP_001139220.1 | 3 - 4 | 327 | 90 | |
GenBank mRNA | BC077053.1 | - | 3 - 4 | 320 | 90 | |
BC161642.1 | - | 3 - 4 | 304 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
