Assay Details
| Gene Symbol: cyp4f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016020.2 | NP_001016020.1 | 13 | 1849 | 94 | |
| XM_018091235.1 | XP_017946724.1 | 13 | 1983 | 94 | ||
| GenBank mRNA | CR848173.2 | - | 13 | 1849 | 94 | |
More Information
Set Membership: |
|

cyp4f2
Genomic Map