Assay Details
| Gene Symbol: lrrc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078852.1 | NP_001072320.1 | 1851 | 102 | ||
| GenBank mRNA | BC121385.1 | - | 1851 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lrrc24| Gene Symbol: lrrc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078852.1 | NP_001072320.1 | 1851 | 102 | ||
| GenBank mRNA | BC121385.1 | - | 1851 | 102 | ||
Set Membership: |
|