Assay Details
| Gene Symbol: LOC100285617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158508.1 | NP_001151980.1 | 198 | 87 | ||
| GenBank mRNA | EU973093.1 | - | 198 | 87 | ||
More Information
Set Membership: |
|

LOC100285617
Genomic Map