Assay Details
| Gene Symbol: hmt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111544.1 | NP_001105014.1 | 452 | 104 | ||
| GenBank mRNA | AF297047.1 | - | 452 | 104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

hmt4| Gene Symbol: hmt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111544.1 | NP_001105014.1 | 452 | 104 | ||
| GenBank mRNA | AF297047.1 | - | 452 | 104 | ||