Assay Details
| Gene Symbol: hmt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111544.1 | NP_001105014.1 | 795 | 64 | ||
| GenBank mRNA | AF297047.1 | - | 795 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hmt4| Gene Symbol: hmt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111544.1 | NP_001105014.1 | 795 | 64 | ||
| GenBank mRNA | AF297047.1 | - | 795 | 64 | ||
Set Membership: |
|