Assay Details
Gene Symbol: sqs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111369.1 | NP_001104839.1 | 430 | 88 | ||
GenBank mRNA | AB007502.1 | - | 430 | 88 | ||
BT067860.1 | - | 440 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
