Assay Details
| Gene Symbol: zfp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112331.1 | NP_001105801.1 | 562 | 103 | ||
| GenBank mRNA | AY599410.1 | - | 562 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

zfp30| Gene Symbol: zfp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112331.1 | NP_001105801.1 | 562 | 103 | ||
| GenBank mRNA | AY599410.1 | - | 562 | 103 | ||
Set Membership: |
|