Assay Details
Gene Symbol: LOC100284744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157639.1 | NP_001151111.1 | 1250 | 92 | ||
GenBank mRNA | BT054895.1 | - | 1233 | 92 | ||
EU969537.1 | - | 1250 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
