Assay Details
| Gene Symbol: pco102443(516) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158518.1 | NP_001151990.1 | 594 | 58 | ||
| GenBank mRNA | EU965494.1 | - | 323 | 58 | ||
| EU973167.1 | - | 594 | 58 | |||
More Information
Set Membership: |
|

pco102443(516)
Genomic Map