Assay Details
| Gene Symbol: umc2551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157818.1 | NP_001151290.1 | 557 | 63 | ||
| GenBank mRNA | EU970134.1 | - | 557 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

umc2551| Gene Symbol: umc2551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157818.1 | NP_001151290.1 | 557 | 63 | ||
| GenBank mRNA | EU970134.1 | - | 557 | 63 | ||
Set Membership: |
|