Assay Details
| Gene Symbol: umc2532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152589.1 | NP_001146061.1 | 1059 | 87 | ||
| GenBank mRNA | BT054529.2 | - | 1059 | 87 | ||
| BT068925.2 | - | 1224 | 87 | |||
More Information
Set Membership: |
|

umc2532
Genomic Map