Assay Details
Gene Symbol: LOC100283642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156543.1 | NP_001150015.1 | 1002 | 77 | ||
GenBank mRNA | BT067338.1 | - | 1129 | 77 | ||
EU965390.1 | - | 1002 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
