Assay Details
| Gene Symbol: lkrsdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111403.1 | NP_001104873.1 | 2159 | 99 | ||
| GenBank mRNA | AF003551.2 | - | 2336 | 99 | ||
| AY110274.1 | - | 2336 | 99 | |||
More Information
Set Membership: |
|

lkrsdh1
Genomic Map