Assay Details
Gene Symbol: TUBB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111847.1 | NP_001105317.1 | 739 | 81 | ||
GenBank mRNA | BT054582.1 | - | 746 | 81 | ||
L10633.1 | - | 739 | 81 |
Gene Symbol: LOC100381281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174180.1 | NP_001167651.1 | 894 | 81 | ||
GenBank mRNA | BT040459.1 | - | 894 | 81 | ||
BT043191.1 | - | 555 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
