Assay Details
| Gene Symbol: adh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111939.1 | NP_001105409.1 | 722 | 71 | ||
| GenBank mRNA | L23548.1 | - | 677 | 71 | ||
| X00580.1 | - | 722 | 71 | |||
More Information
Set Membership: |
|

adh1
Genomic Map