Assay Details
| Gene Symbol: adh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111939.1 | NP_001105409.1 | 962 | 60 | ||
| GenBank mRNA | J01239.1 | - | 230 | 60 | ||
| X00580.1 | - | 962 | 60 | |||
More Information
Set Membership: |
|

adh1
Genomic Map