Assay Details
| Gene Symbol: mir1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112101.1 | NP_001105571.1 | 808 | 73 | ||
| GenBank mRNA | AF019145.2 | - | 808 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mir1| Gene Symbol: mir1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112101.1 | NP_001105571.1 | 808 | 73 | ||
| GenBank mRNA | AF019145.2 | - | 808 | 73 | ||
Set Membership: |
|