Assay Details
Gene Symbol: GIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112561.1 | NP_001106031.1 | 317 | 64 | ||
GenBank mRNA | BT054882.1 | - | 357 | 64 | ||
EF515855.1 | - | 317 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
