Assay Details
| Gene Symbol: CFM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112591.1 | NP_001106061.1 | 1195 | 82 | ||
| GenBank mRNA | EU084957.1 | - | 1195 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFM3| Gene Symbol: CFM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112591.1 | NP_001106061.1 | 1195 | 82 | ||
| GenBank mRNA | EU084957.1 | - | 1195 | 82 | ||