Assay Details
| Gene Symbol: CPNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112246.1 | NP_001105716.1 | 2047 | 105 | ||
| GenBank mRNA | Z12114.1 | - | 2047 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPNA| Gene Symbol: CPNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112246.1 | NP_001105716.1 | 2047 | 105 | ||
| GenBank mRNA | Z12114.1 | - | 2047 | 105 | ||
Set Membership: |
|