Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| FARS2 - phenylalanyl-tRNA synthetase 2, mitochondrial | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001318872.1 | Intron | NP_001305801.1 | ||||
| NM_006567.4 | Intron | NP_006558.1 | ||||
| XM_005248812.3 | Intron | XP_005248869.1 | ||||
| XM_006714966.2 | Intron | XP_006715029.1 | ||||
| XM_011514247.2 | Intron | XP_011512549.1 | ||||
| XM_011514248.2 | Intron | XP_011512550.1 | ||||
| XM_011514249.1 | Intron | XP_011512551.1 | ||||
| XM_011514251.2 | Intron | XP_011512553.1 | ||||
| XM_017010186.1 | Intron | XP_016865675.1 | ||||
| XM_017010187.1 | Intron | XP_016865676.1 | ||||
| LOC101927950 - uncharacterized LOC101927950 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

FARS2
Genomic Map