Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR371A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| MIR371A - microRNA 371a | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MIR371B - microRNA 371b | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MIR372 - microRNA 372 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MIR373 - microRNA 373 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| NLRP12 - NLR family pyrin domain containing 12 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001277126.1 | Intron | NP_001264055.1 | ||||
| NM_001277129.1 | Intron | NP_001264058.1 | ||||
| NM_144687.3 | Intron | NP_653288.1 | ||||
| XM_011527479.1 | Intron | XP_011525781.1 | ||||
| XM_011527480.1 | Intron | XP_011525782.1 | ||||
| XM_011527482.1 | Intron | XP_011525784.1 | ||||
| XM_017027460.1 | Intron | XP_016882949.1 | ||||
| XM_017027461.1 | Intron | XP_016882950.1 | ||||
| XM_017027462.1 | Intron | XP_016882951.1 | ||||
| XM_017027463.1 | Intron | XP_016882952.1 | ||||
| XM_017027464.1 | Intron | XP_016882953.1 | ||||
| XM_017027465.1 | Intron | XP_016882954.1 | ||||
| XM_017027466.1 | Intron | XP_016882955.1 | ||||
| XM_017027467.1 | Intron | XP_016882956.1 | ||||

MIR371A
Genomic Map